Assay Details
| Gene Symbol: NERD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335477.1 | NP_001324854.1 | 1 | 2016 | 73 | |
| NM_127202.5 | NP_179241.4 | 1 | 1937 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NERD| Gene Symbol: NERD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335477.1 | NP_001324854.1 | 1 | 2016 | 73 | |
| NM_127202.5 | NP_179241.4 | 1 | 1937 | 73 | ||
Set Membership: |
|