Assay Details
| Gene Symbol: ROC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084432.1 | NP_001077901.1 | 2 | 623 | 112 | |
| NM_001335481.1 | NP_001318231.1 | 1 | 1073 | 112 | ||
| GenBank mRNA | AY048215.1 | - | 1 | 688 | 112 | |
| U40399.1 | - | 1 | 682 | 112 | ||
More Information
Set Membership: |
|

ROC3
Genomic Map