Assay Details
| Gene Symbol: AT2G16730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127225.3 | NP_179264.2 | 10 - 11 | 1384 | 102 | |
| GenBank mRNA | AJ270309.1 | - | 10 - 11 | 1205 | 102 | |
More Information
Set Membership: |
|

AT2G16730
Genomic Map