Assay Details
| Gene Symbol: Kin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127276.3 | NP_565408.1 | 2 - 3 | 838 | 69 | |
| NM_201749.1 | NP_973478.1 | 2 - 3 | 812 | 69 | ||
| GenBank mRNA | AF370585.1 | - | 2 - 3 | 407 | 69 | |
| AY049263.1 | - | 2 - 3 | 806 | 69 | ||
| AY149966.1 | - | 2 - 3 | 407 | 69 | ||
More Information
Set Membership: |
|

Kin3
Genomic Map