Assay Details
| Gene Symbol: EMB2763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202612.1 | NP_001189541.1 | 24 - 25 | 2945 | 100 | |
| NM_127305.3 | NP_565418.1 | 23 - 24 | 2727 | 100 | ||
| GenBank mRNA | AY057479.1 | - | 23 - 24 | 2693 | 100 | |
| AY090298.1 | - | 23 - 24 | 2608 | 100 | ||
More Information
Set Membership: |
|

EMB2763
Genomic Map