Assay Details
| Gene Symbol: EMB2763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202612.1 | NP_001189541.1 | 7 - 8 | 888 | 114 | |
| NM_127305.3 | NP_565418.1 | 6 - 7 | 772 | 114 | ||
| GenBank mRNA | AY057479.1 | - | 6 - 7 | 738 | 114 | |
| AY090298.1 | - | 6 - 7 | 653 | 114 | ||
More Information
Set Membership: |
|

EMB2763
Genomic Map