Assay Details
| Gene Symbol: HMGB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036287.3 | NP_001031364.2 | 1 - 2 | 181 | 70 | |
| NM_001084440.1 | NP_001077909.1 | 2 - 3 | 109 | 70 | ||
| NM_127310.3 | NP_179347.1 | 2 - 3 | 211 | 70 | ||
| GenBank mRNA | AF370562.1 | - | 1 - 2 | 44 | 70 | |
| AK317044.1 | - | 2 - 3 | 80 | 70 | ||
| AY150467.1 | - | 1 - 2 | 44 | 70 | ||
More Information
Set Membership: |
|

HMGB4
Genomic Map