Assay Details
| Gene Symbol: HMGB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036287.3 | NP_001031364.2 | 6 - 7 | 537 | 113 | |
| NM_127310.3 | NP_179347.1 | 7 - 8 | 567 | 113 | ||
| GenBank mRNA | AF370562.1 | - | 6 - 7 | 400 | 113 | |
| AY150467.1 | - | 6 - 7 | 400 | 113 | ||
More Information
Set Membership: |
|

HMGB4
Genomic Map