Assay Details
| Gene Symbol: ATSERAT3;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127318.5 | NP_565421.1 | 8 - 9 | 1130 | 77 | |
| GenBank mRNA | AF112303.1 | - | 8 - 9 | 884 | 77 | |
| AK176573.1 | - | 8 - 9 | 877 | 77 | ||
| BT024717.1 | - | 8 - 9 | 861 | 77 | ||
More Information
Set Membership: |
|

ATSERAT3;1
Genomic Map