Assay Details
| Gene Symbol: STY8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127324.5 | NP_179361.1 | 3 - 4 | 566 | 78 | |
| GenBank mRNA | AY070086.1 | - | 3 - 4 | 516 | 78 | |
| AY096470.1 | - | 3 - 4 | 399 | 78 | ||
More Information
Set Membership: |
|

STY8
Genomic Map