Assay Details
| Gene Symbol: AT2G17710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127325.2 | NP_565423.1 | 2 - 3 | 567 | 66 | |
| GenBank mRNA | AF446349.1 | - | 2 - 3 | 419 | 66 | |
| AK228455.1 | - | 2 - 3 | 325 | 66 | ||
| AY097418.1 | - | 2 - 3 | 307 | 66 | ||
More Information
Set Membership: |
|

AT2G17710
Genomic Map