Assay Details
| Gene Symbol: NIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202615.1 | NP_001189544.1 | 2 - 3 | 539 | 111 | |
| NM_127327.4 | NP_179364.2 | 2 - 3 | 681 | 111 | ||
| GenBank mRNA | AJ400898.1 | - | 2 - 3 | 400 | 111 | |
More Information
Set Membership: |
|

NIP2
Genomic Map