Assay Details
| Gene Symbol: VPS35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127333.5 | NP_179370.2 | 16 - 17 | 1875 | 97 | |
| GenBank mRNA | BT008553.1 | - | 16 - 17 | 1753 | 97 | |
| BT008664.1 | - | 16 - 17 | 1803 | 97 | ||
More Information
Set Membership: |
|

VPS35A
Genomic Map