Assay Details
| Gene Symbol: VPS35A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127333.5 | NP_179370.2 | 17 - 18 | 1959 | 95 | |
| GenBank mRNA | BT008553.1 | - | 17 - 18 | 1837 | 95 | |
| BT008664.1 | - | 17 - 18 | 1887 | 95 | ||
More Information
Set Membership: |
|

VPS35A
Genomic Map