Assay Details
| Gene Symbol: AT2G41835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180027.3 | NP_850358.1 | 1 - 2 | 375 | 111 | |
| GenBank mRNA | AY065272.1 | - | 1 - 2 | 238 | 111 | |
| AY096620.1 | - | 1 - 2 | 98 | 111 | ||
More Information
Set Membership: |
|

AT2G41835
Genomic Map