Assay Details
| Gene Symbol: POP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202810.1 | NP_001189739.1 | 5 - 6 | 550 | 77 | |
| NM_001337019.1 | NP_001324792.1 | 4 - 5 | 676 | 77 | ||
| NM_129881.3 | NP_565993.1 | 4 - 5 | 686 | 77 | ||
| NM_180058.2 | NP_850389.1 | 3 - 4 | 783 | 77 | ||
| NM_201949.2 | NP_973678.1 | 4 - 5 | 683 | 77 | ||
| GenBank mRNA | AY086048.1 | - | 4 - 5 | 655 | 77 | |
More Information
Set Membership: |
|

POP4
Genomic Map