Assay Details
| Gene Symbol: LARP6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129959.3 | NP_566004.1 | 2 - 3 | 872 | 136 | |
| NM_180075.2 | NP_850406.1 | 2 - 3 | 872 | 136 | ||
| GenBank mRNA | AF332565.1 | - | 2 - 3 | 685 | 136 | |
| AF367277.1 | - | 2 - 3 | 872 | 136 | ||
| AF375410.1 | - | 2 - 3 | 811 | 136 | ||
| AY056238.1 | - | 2 - 3 | 872 | 136 | ||
| AY129474.1 | - | 2 - 3 | 685 | 136 | ||
More Information
Set Membership: |
|

LARP6b
Genomic Map