Assay Details
| Gene Symbol: BGLU28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337087.1 | NP_001324080.1 | 5 - 6 | 753 | 92 | |
| NM_180085.4 | NP_850416.1 | 10 - 11 | 1328 | 92 | ||
| GenBank mRNA | BT023443.1 | - | 10 - 11 | 1167 | 92 | |
More Information
Set Membership: |
|

BGLU28
Genomic Map