Assay Details
| Gene Symbol: BGLU28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337087.1 | NP_001324080.1 | 3 - 4 | 497 | 102 | |
| NM_180085.4 | NP_850416.1 | 8 - 9 | 1072 | 102 | ||
| GenBank mRNA | BT023443.1 | - | 8 - 9 | 911 | 102 | |
More Information
Set Membership: |
|

BGLU28
Genomic Map