Assay Details
| Gene Symbol: BGLU28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337087.1 | NP_001324080.1 | 4 - 5 | 722 | 66 | |
| NM_180085.4 | NP_850416.1 | 9 - 10 | 1297 | 66 | ||
| GenBank mRNA | BT023443.1 | - | 9 - 10 | 1136 | 66 | |
More Information
Set Membership: |
|

BGLU28
Genomic Map