Assay Details
| Gene Symbol: BGLU29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084587.1 | NP_001078056.1 | 9 - 10 | 1182 | 71 | |
| NM_001125052.1 | NP_001118524.1 | 9 - 10 | 1182 | 71 | ||
| NM_180086.1 | NP_850417.1 | 9 - 10 | 1182 | 71 | ||
| GenBank mRNA | AK118055.1 | - | 9 - 10 | 1182 | 71 | |
More Information
Set Membership: |
|

BGLU29
Genomic Map