Assay Details
| Gene Symbol: DL1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180087.1 | NP_850418.1 | 1 - 2 | 133 | 109 | |
| NM_180088.3 | NP_850419.1 | 1 - 2 | 267 | 109 | ||
| NM_180089.2 | NP_850420.1 | 1 - 2 | 275 | 109 | ||
| GenBank mRNA | AF488807.1 | - | 1 - 2 | 133 | 109 | |
| AJ304843.1 | - | 1 - 2 | 267 | 109 | ||
| AJ304844.1 | - | 1 - 2 | 267 | 109 | ||
| BT046114.1 | - | 1 - 2 | 133 | 109 | ||
More Information
Set Membership: |
|

DL1D
Genomic Map