Assay Details
| Gene Symbol: DL1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180087.1 | NP_850418.1 | 9 - 10 | 942 | 99 | |
| NM_180088.3 | NP_850419.1 | 9 - 10 | 1076 | 99 | ||
| NM_180089.2 | NP_850420.1 | 10 - 11 | 1135 | 99 | ||
| GenBank mRNA | AF488807.1 | - | 9 - 10 | 942 | 99 | |
| AJ304843.1 | - | 10 - 11 | 1127 | 99 | ||
| AJ304844.1 | - | 9 - 10 | 1076 | 99 | ||
| BT046114.1 | - | 10 - 11 | 993 | 99 | ||
More Information
Set Membership: |
|

DL1D
Genomic Map