Assay Details
| Gene Symbol: GC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180116.2 | NP_850447.1 | 4 - 5 | 708 | 76 | |
| GenBank mRNA | AY069912.1 | - | 4 - 5 | 649 | 76 | |
| BT001025.1 | - | 4 - 5 | 559 | 76 | ||
| EU249331.1 | - | 4 - 5 | 559 | 76 | ||
More Information
Set Membership: |
|

GC4
Genomic Map