Assay Details
| Gene Symbol: ROPGAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180127.2 | NP_850458.1 | 1 - 2 | 1290 | 87 | |
| GenBank mRNA | AK117311.1 | - | 1 - 2 | 500 | 87 | |
| BT005369.1 | - | 1 - 2 | 432 | 87 | ||
More Information
Set Membership: |
|

ROPGAP3
Genomic Map