Assay Details
| Gene Symbol: CCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337221.1 | NP_001318437.1 | 3 - 4 | 389 | 120 | |
| NM_180129.4 | NP_850460.1 | 3 - 4 | 482 | 120 | ||
| GenBank mRNA | AY519511.1 | - | 2 - 3 | 152 | 120 | |
| U28422.1 | - | 3 - 4 | 389 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001337221.1 transcripts.
Set Membership: |
|

CCA1
Genomic Map