Assay Details
| Gene Symbol: BOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084602.2 | NP_001078071.1 | 10 - 11 | 2294 | 66 | |
| NM_180138.3 | NP_850469.1 | 11 - 12 | 2385 | 66 | ||
| GenBank mRNA | AB073713.1 | - | 11 - 12 | 1844 | 66 | |
| AY070067.1 | - | 11 - 12 | 2200 | 66 | ||
| AY096436.1 | - | 11 - 12 | 1844 | 66 | ||
| BT000732.1 | - | 11 - 12 | 2200 | 66 | ||
More Information
Set Membership: |
|

BOR1
Genomic Map