Assay Details
| Gene Symbol: BOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180138.3 | NP_850469.1 | 2 - 3 | 726 | 67 | |
| GenBank mRNA | AB073713.1 | - | 2 - 3 | 185 | 67 | |
| AY070067.1 | - | 2 - 3 | 541 | 67 | ||
| AY096436.1 | - | 2 - 3 | 185 | 67 | ||
| BT000732.1 | - | 2 - 3 | 541 | 67 | ||
More Information
Set Membership: |
|

BOR1
Genomic Map