Assay Details
| Gene Symbol: BOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084602.2 | NP_001078071.1 | 7 - 8 | 1573 | 64 | |
| NM_180138.3 | NP_850469.1 | 8 - 9 | 1664 | 64 | ||
| GenBank mRNA | AB073713.1 | - | 8 - 9 | 1123 | 64 | |
| AY070067.1 | - | 8 - 9 | 1479 | 64 | ||
| AY096436.1 | - | 8 - 9 | 1123 | 64 | ||
| BT000732.1 | - | 8 - 9 | 1479 | 64 | ||
More Information
Set Membership: |
|

BOR1
Genomic Map