Assay Details
| Gene Symbol: BOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084602.2 | NP_001078071.1 | 8 - 9 | 1700 | 71 | |
| NM_180138.3 | NP_850469.1 | 9 - 10 | 1791 | 71 | ||
| GenBank mRNA | AB073713.1 | - | 9 - 10 | 1250 | 71 | |
| AY070067.1 | - | 9 - 10 | 1606 | 71 | ||
| AY096436.1 | - | 9 - 10 | 1250 | 71 | ||
| BT000732.1 | - | 9 - 10 | 1606 | 71 | ||
More Information
Set Membership: |
|

BOR1
Genomic Map