Assay Details
| Gene Symbol: SWI3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337268.1 | NP_001324817.1 | 1 - 2 | 150 | 77 | |
| NM_180145.3 | NP_850476.1 | 1 - 2 | 142 | 77 | ||
| GenBank mRNA | AY062794.1 | - | 1 - 2 | 121 | 77 | |
| AY081570.1 | - | 1 - 2 | 55 | 77 | ||
More Information
Set Membership: |
|

SWI3A
Genomic Map