Assay Details
| Gene Symbol: RFI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337272.1 | NP_001324431.1 | 4 - 5 | 675 | 69 | |
| NM_180147.3 | NP_850478.1 | 4 - 5 | 738 | 69 | ||
| GenBank mRNA | BT002412.1 | - | 4 - 5 | 641 | 69 | |
| BT008845.1 | - | 4 - 5 | 442 | 69 | ||
| DQ086859.1 | - | 4 - 5 | 442 | 69 | ||
More Information
Set Membership: |
|

RFI2
Genomic Map