Assay Details
| Gene Symbol: RFI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180147.3 | NP_850478.1 | 5 - 6 | 1037 | 86 | |
| GenBank mRNA | BT002412.1 | - | 5 - 6 | 940 | 86 | |
| BT008845.1 | - | 5 - 6 | 741 | 86 | ||
| DQ086859.1 | - | 5 - 6 | 741 | 86 | ||
More Information
Set Membership: |
|

RFI2
Genomic Map