Assay Details
| Gene Symbol: KIN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125074.2 | NP_001118546.1 | 7 - 8 | 1123 | 88 | |
| NM_110974.5 | NP_566130.1 | 7 - 8 | 1239 | 88 | ||
| NM_180157.1 | NP_850488.1 | 7 - 8 | 1381 | 88 | ||
| GenBank mRNA | AY093170.1 | - | 7 - 8 | 1381 | 88 | |
| BT010386.1 | - | 7 - 8 | 1037 | 88 | ||
| DQ778957.1 | - | 6 - 7 | 968 | 88 | ||
| X94757.1 | - | 1097 | 88 | |||
More Information
Set Membership: |
|

KIN10
Genomic Map