Assay Details
| Gene Symbol: MS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125092.1 | NP_001118564.1 | 5 - 6 | 1169 | 83 | |
| NM_111249.5 | NP_187028.1 | 6 - 7 | 1323 | 83 | ||
| NM_180176.4 | NP_850507.1 | 6 - 7 | 1419 | 83 | ||
| GenBank mRNA | AJ608674.1 | - | 5 - 6 | 1126 | 83 | |
| AY084559.1 | - | 6 - 7 | 1200 | 83 | ||
| AY150385.1 | - | 5 - 6 | 1121 | 83 | ||
More Information
Set Membership: |
|

MS2
Genomic Map