Assay Details
| Gene Symbol: AT3G03870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180177.2 | NP_850508.1 | 1 | 956 | 111 | |
| GenBank mRNA | AY080808.1 | - | 1 | 874 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G03870| Gene Symbol: AT3G03870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180177.2 | NP_850508.1 | 1 | 956 | 111 | |
| GenBank mRNA | AY080808.1 | - | 1 | 874 | 111 | |
Set Membership: |
|