Assay Details
| Gene Symbol: SYP43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111445.5 | NP_566256.1 | 3 - 4 | 908 | 93 | |
| NM_180188.3 | NP_850519.1 | 3 - 4 | 854 | 93 | ||
| GenBank mRNA | AJ245408.1 | - | 3 - 4 | 830 | 93 | |
| AY099723.1 | - | 3 - 4 | 854 | 93 | ||
More Information
Set Membership: |
|

SYP43
Genomic Map