Assay Details
| Gene Symbol: SYP43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111445.5 | NP_566256.1 | 4 - 5 | 974 | 145 | |
| NM_180188.3 | NP_850519.1 | 4 - 5 | 920 | 145 | ||
| GenBank mRNA | AJ245408.1 | - | 4 - 5 | 896 | 145 | |
| AY099723.1 | - | 4 - 5 | 920 | 145 | ||
More Information
Set Membership: |
|

SYP43
Genomic Map