Assay Details
| Gene Symbol: IMPA-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111552.3 | NP_187328.1 | 3 - 4 | 630 | 88 | |
| NM_180193.3 | NP_850524.1 | 3 - 4 | 610 | 88 | ||
| GenBank mRNA | AF077528.1 | - | 3 - 4 | 443 | 88 | |
| AY088732.1 | - | 3 - 4 | 632 | 88 | ||
| AY094390.1 | - | 3 - 4 | 456 | 88 | ||
| AY125529.1 | - | 3 - 4 | 395 | 88 | ||
More Information
Set Membership: |
|

IMPA-1
Genomic Map