Assay Details
| Gene Symbol: IMPA-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111552.3 | NP_187328.1 | 9 - 10 | 1457 | 81 | |
| NM_180193.3 | NP_850524.1 | 9 - 10 | 1437 | 81 | ||
| GenBank mRNA | AF077528.1 | - | 9 - 10 | 1270 | 81 | |
| AY088732.1 | - | 9 - 10 | 1459 | 81 | ||
| AY094390.1 | - | 9 - 10 | 1283 | 81 | ||
| AY125529.1 | - | 9 - 10 | 1222 | 81 | ||
More Information
Set Membership: |
|

IMPA-1
Genomic Map