Assay Details
| Gene Symbol: BCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111565.6 | NP_187341.1 | 3 - 4 | 603 | 82 | |
| NM_180196.3 | NP_850527.1 | 4 - 5 | 518 | 82 | ||
| GenBank mRNA | AK316767.1 | - | 3 - 4 | 501 | 82 | |
| AK317408.1 | - | 4 - 5 | 463 | 82 | ||
| AY086441.1 | - | 3 - 4 | 500 | 82 | ||
More Information
Set Membership: |
|

BCE2
Genomic Map