Assay Details
| Gene Symbol: iPGAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111693.3 | NP_187471.1 | 6 - 7 | 1496 | 87 | |
| NM_180211.2 | NP_850542.1 | 5 - 6 | 1442 | 87 | ||
| GenBank mRNA | AF424615.1 | - | 6 - 7 | 1357 | 87 | |
| AK316907.1 | - | 5 - 6 | 1443 | 87 | ||
| AY039969.1 | - | 6 - 7 | 1358 | 87 | ||
| AY113910.1 | - | 5 - 6 | 1268 | 87 | ||
More Information
Set Membership: |
|

iPGAM2
Genomic Map