Assay Details
| Gene Symbol: LSM1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338100.1 | NP_001326378.1 | 3 - 4 | 377 | 154 | |
| NM_112264.4 | NP_566476.1 | 3 - 4 | 434 | 154 | ||
| NM_180249.4 | NP_850580.1 | 2 - 3 | 544 | 154 | ||
| GenBank mRNA | AY084768.1 | - | 3 - 4 | 377 | 154 | |
| BT002795.1 | - | 2 - 3 | 466 | 154 | ||
| BT004340.1 | - | 2 - 3 | 135 | 154 | ||
More Information
Set Membership: |
|

LSM1B
Genomic Map