Assay Details
| Gene Symbol: LSM1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338100.1 | NP_001326378.1 | 5 - 6 | 539 | 64 | |
| NM_001338101.1 | NP_001326377.1 | 3 - 4 | 295 | 64 | ||
| NM_112264.4 | NP_566476.1 | 5 - 6 | 604 | 64 | ||
| NM_180249.4 | NP_850580.1 | 4 - 5 | 714 | 64 | ||
| GenBank mRNA | AY084768.1 | - | 5 - 6 | 547 | 64 | |
| BT002795.1 | - | 4 - 5 | 636 | 64 | ||
| BT004340.1 | - | 4 - 5 | 305 | 64 | ||
More Information
Set Membership: |
|

LSM1B
Genomic Map