Assay Details
| Gene Symbol: GRP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113288.3 | NP_189025.1 | 3 - 4 | 528 | 90 | |
| NM_180298.4 | NP_850629.1 | 3 - 4 | 516 | 90 | ||
| GenBank mRNA | AY070755.1 | - | 3 - 4 | 285 | 90 | |
| AY097374.1 | - | 2 - 3 | 190 | 90 | ||
More Information
Set Membership: |
|

GRP4
Genomic Map