Assay Details
| Gene Symbol: MSH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338685.1 | NP_001326470.1 | 2 - 3 | 1067 | 111 | |
| NM_180299.1 | NP_850630.1 | 2 - 3 | 925 | 111 | ||
More Information
Set Membership: |
|

MSH7
Genomic Map