Assay Details
Gene Symbol: MSH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338685.1 | NP_001326470.1 | 9 - 10 | 1605 | 72 | |
NM_180299.1 | NP_850630.1 | 9 - 10 | 1463 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
