Assay Details
| Gene Symbol: DCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_099986.4 | NP_171612.1 | 12 - 13 | 3457 | 85 | |
| GenBank mRNA | AF292940.1 | - | 12 - 13 | 3432 | 85 | |
More Information
Set Membership: |
|

DCL1
Genomic Map