Assay Details
| Gene Symbol: DCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197952.1 | NP_001184881.1 | 1 - 2 | 1037 | 108 | |
| NM_099986.4 | NP_171612.1 | 1 - 2 | 1332 | 108 | ||
| GenBank mRNA | AF292940.1 | - | 1 - 2 | 1307 | 108 | |
More Information
Set Membership: |
|

DCL1
Genomic Map