Assay Details
| Gene Symbol: DCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197952.1 | NP_001184881.1 | 9 - 10 | 2665 | 78 | |
| NM_099986.4 | NP_171612.1 | 9 - 10 | 2960 | 78 | ||
| GenBank mRNA | AF292940.1 | - | 9 - 10 | 2935 | 78 | |
More Information
Set Membership: |
|

DCL1
Genomic Map